missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Nicotinate, Mikromol™

Product Code. 17940476
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
This item is not returnable. View return policy

Product Code. 17940476

Brand: Mikromol MM1756.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Ethyl Nicotinate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl Nicotinate
CAS 614-18-6
Analyte or Component Names Ethyl Nicotinate
Shipping Condition Room Temperature
Molecular Formula C8 H9 N O2
Certifications/Compliance ISO 17025
Synonym Ethyl Nicotinate, 3-Pyridinecarboxylic acid, ethyl ester, Nicotinic acid, ethyl ester (6CI,8CI), 3-(Ethoxycarbonyl)pyridine, 3-Carbethoxypyridine, Ba 2673, Ethyl 3-pyridinecarboxylate, Ethyl nicotinate, Ignicut, Ignocut, Mucotherm, NSC 8872, Nicaethan, Nikethan, Nikithan, beta-Pyridinecarboxylic acid ethyl ester
InChI Formula InChI=1S/C8H9NO2/c1-2-11-8(10)7-4-3-5-9-6-7/h3-6H,2H2,1H3
SMILES CCOC(=O)c1cccnc1
IUPAC Name ethyl pyridine-3-carboxylate
Molecular Weight (g/mol) 151.16
Quantity 250 mg
Formula Weight 151.0633 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.