missing translation for 'onlineSavingsMsg'
Learn More

Ethyl nicotinate, 99%

Product Code. 11312066
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 11312066

Brand: Thermo Scientific Alfa Aesar L03215.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 614-18-6 | C8H9NO2 | 151.165 g/mol

Ethyl nicotinate is used for skin-conditioning cosmetics. It is principally used in medicine as a local vasodilator at a concentration of 1 to 2 % in creams, ointments and pomades. It is utilized for the treatment sprains, wrenches, muscular pains and tendonitis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl nicotinate is used for skin-conditioning cosmetics. It is principally used in medicine as a local vasodilator at a concentration of 1 to 2 % in creams, ointments and pomades. It is utilized for the treatment sprains, wrenches, muscular pains and tendonitis.

Solubility
Miscible with water.

Notes
Air and light sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 614-18-6
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD00006389
InChI Key XBLVHTDFJBKJLG-UHFFFAOYSA-N
Synonym ethyl nicotinate, ethyl nicotinoate, nicotinic acid ethyl ester, ethyl 3-pyridinecarboxylate, 3-pyridinecarboxylic acid, ethyl ester, ethylnicotinate, nicotinic acid, ethyl ester, mucotherm, nicaethan, nikethan
PubChem CID 69188
IUPAC Name ethyl pyridine-3-carboxylate
SMILES CCOC(=O)C1=CN=CC=C1

Specifications

Melting Point 8°C to 9°C
Density 1.107
Boiling Point 223°C to 224°C
Flash Point 93°C (199°F)
Odor Characteristic
Refractive Index 1.503
Quantity 100 g
Beilstein 122937
Solubility Information Miscible with water.
Formula Weight 151.17
Percent Purity 99%
Chemical Name or Material Ethyl nicotinate
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl nicotinate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Ethyl nicotinate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.