missing translation for 'onlineSavingsMsg'
Learn More

Ethyl N-phenylformimidate, 99%, Thermo Scientific™

Product Code. 10773251 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10773251 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10773251 Supplier Thermo Scientific Acros Supplier No. 278750250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 6780-49-0
Color Colorless to Yellow
Boiling Point 112.0°C to 114.0°C (33.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C9H11NO
Refractive Index 1.5260 to 1.5280
MDL Number MFCD00043655
Quantity 25 g
Synonym ethyl n-phenylformimidate, ethyl isoformanilide, n-ethoxymethyleneaniline, methanimidic acid, n-phenyl-, ethyl ester, ethyl phenylimidoformate, n-ethoxymethylene aniline, ethyl n-phenylcarboximidate, ethylisoformanilide, e-ethyl n-phenylcarboximidate, ethyl iso-formanilide
InChI Key DRDBNKYFCOLNQO-UHFFFAOYSA-N
SMILES CCOC=NC1=CC=CC=C1
IUPAC Name ethyl N-phenylcarboximidate
Molecular Weight (g/mol) 149.19
PubChem CID 111236
Formula Weight 149.19
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Ethyl N-phenylformimidate, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl N-phenylformimidate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.