missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl N-n-Butyl-d9-carbamate, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.10g
0.25g
Specifications
Specifications
| Chemical Name or Material | Ethyl N-Butylcarbamate-D9 (butyl-D9) |
| CAS | 1276197-24-0 |
| Molecular Formula | C7 D9 H6 N O2 |
| Quantity | 0.1 g |
| Synonym | Ethyl N-n-Butyl-d9-carbamate, Ethyl butyl-d9-carbamate, Ethyl N-butylcarbamate-D9 (butyl-D9), N-(Butyl-d9)-O-ethylurethane, N-(Butyl-d9)urethane, N-Butylcarbamic-d9 acid ethyl ester |
| InChI Formula | InChI=1 S/C7H15NO2/c1-3-5-6-8-7(9)10-4-2/h3-6H2,1-2H3,(H,8,9)/i1D3,3D2,5D2,6D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])NC(=O)OCC |
| IUPAC Name | ethyl N-(1,1,2,2,3,3,4,4,4-nonadeuteriobutyl)carbamate |
| Molecular Weight (g/mol) | 154.255 |
| Formula Weight | 154.1668 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction