missing translation for 'onlineSavingsMsg'
Learn More

Ethyl N-(diphenylmethylene)glycinate, 98%

Product Code. 10376620 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10376620 5 g Glass bottle 5g
10627484 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10376620 Supplier Thermo Scientific Supplier No. 124130050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 69555-14-2 | C17H17NO2 | 267.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69555-14-2
Molecular Formula C17H17NO2
Molecular Weight (g/mol) 267.32
MDL Number MFCD00010590
InChI Key QUGJYNGNUBHTNS-UHFFFAOYSA-N
Synonym ethyl n-diphenylmethylene glycinate, n-diphenylmethylene glycine ethyl ester, ethyl 2-diphenylmethylene amino acetate, ethyl 2-diphenylmethyleneamino acetate, diphenylmethylene-glycine ethyl ester, ethyl 2-benzhydrylideneamino acetate, ethyl 2-diphenylmethylidene amino acetate, diphenylmethylene glycine ethylester, ethyl diphenylmethylenamino acetate, diphenylmethylene glycine ethyl ester
PubChem CID 319508
IUPAC Name ethyl 2-(benzhydrylideneamino)acetate
SMILES CCOC(=O)CN=C(C1=CC=CC=C1)C2=CC=CC=C2

Specifications

Melting Point 51°C to 55°C
Color White to Yellow
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (C6H5)2C=NCH2COOCH2CH3
Quantity 5 g
Formula Weight 267.32
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Ethyl N-(diphenylmethylene)glycinate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.