missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ethyl N-(4-bromophenyl)oxamate

Product Code. p-7033478
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11430090

Brand: Alfa Aesar™ H50598.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 24451-15-8
Molecular Formula C10H10BrNO3
Molecular Weight (g/mol) 272.098
MDL Number MFCD00728542
InChI Key RWWFLEVLFXJHIB-UHFFFAOYSA-N
Synonym ethyl 4-bromoanilino oxo acetate, ethyl 4-bromophenyl carbamoyl formate, ethyl 2-4-bromophenylamino-2-oxoacetate, ethyl n-p-bromophenyloxamate, ethyl 4-bromophenyl amino oxo acetate, ethyl 2-4-bromoanilino-2-oxoacetate, ethyl n-4-bromophenyl carbamoyl formate, ethyl n-4-bromophenyl oxamate, n-4-bromophenyl oxamidic acid ethyl ester, ethyl 2-4-bromophenyl amino-2-oxoacetate
PubChem CID 2794843
IUPAC Name ethyl 2-(4-bromoanilino)-2-oxoacetate
SMILES CCOC(=O)C(=O)NC1=CC=C(C=C1)Br

Specifications

Quantity 1 g
Formula Weight 272.1
Chemical Name or Material Ethyl N-(4-bromophenyl)oxamate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.