missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ethyl N-(4-bromophenyl)oxamate

Product Code. 11440090
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11440090 5 g 5g
11430090 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11440090 Supplier Alfa Aesar™ Supplier No. H50598.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 24451-15-8
Molecular Formula C10H10BrNO3
Molecular Weight (g/mol) 272.098
MDL Number MFCD00728542
InChI Key RWWFLEVLFXJHIB-UHFFFAOYSA-N
Synonym ethyl 4-bromoanilino oxo acetate, ethyl 4-bromophenyl carbamoyl formate, ethyl 2-4-bromophenylamino-2-oxoacetate, ethyl n-p-bromophenyloxamate, ethyl 4-bromophenyl amino oxo acetate, ethyl 2-4-bromoanilino-2-oxoacetate, ethyl n-4-bromophenyl carbamoyl formate, ethyl n-4-bromophenyl oxamate, n-4-bromophenyl oxamidic acid ethyl ester, ethyl 2-4-bromophenyl amino-2-oxoacetate
PubChem CID 2794843
IUPAC Name ethyl 2-(4-bromoanilino)-2-oxoacetate
SMILES CCOC(=O)C(=O)NC1=CC=C(C=C1)Br

Specifications

Quantity 5 g
Formula Weight 272.1
Chemical Name or Material Ethyl N-(4-bromophenyl)oxamate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.