missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Myristate, TRC
Specifications
Specifications
| Chemical Name or Material | Tetradecanoic acid ethyl ester |
| CAS | 124-06-1 |
| Molecular Formula | C16 H32 O2 |
| Quantity | 10 g |
| Synonym | Tetradecanoic acid, ethyl ester (9CI, ACI), Myristic acid, ethyl ester (6CI, 7CI, 8CI), Ethyl myristate, Ethyl tetradecanoate, Nikkol BM, NSC 8917 |
| InChI Formula | InChI=1S/C16H32O2/c1-3-5-6-7-8-9-10-11-12-13-14-15-16(17)18-4-2/h3-15H2,1-2H3 |
| SMILES | CCCCCCCCCCCCCC(=O)OCC |
| IUPAC Name | ethyl tetradecanoate |
| Molecular Weight (g/mol) | 256.42 |
| Formula Weight | 256.2402 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?