missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Myristate, TRC

Product Code. 30384097
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30384097 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30384097 Supplier Toronto Research Chemicals Supplier No. TRCE9251801G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethylpalmitate
CAS 628-97-7
Molecular Formula C18 H36 O2
Quantity 1 g
Synonym Hexadecanoic acid, ethyl ester (9CI, ACI), Palmitic acid, ethyl ester (6CI, 8CI), Ethyl hexadecanoate, Ethyl palmitate, NSC 8918, Hexadecanoic acid ethyl ester, Palmitic acid ethyl ester
InChI Formula InChI=1S/C18H36O2/c1-3-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20-4-2/h3-17H2,1-2H3
SMILES CCCCCCCCCCCCCCCC(=O)OCC
IUPAC Name ethyl hexadecanoate
Molecular Weight (g/mol) 284.48
Formula Weight 284.2715
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.