missing translation for 'onlineSavingsMsg'
Learn More

Ethyl myristate, +98%, ACROS Organics™

Brand:  Acros Organics 385540250

Code : 29

Additional Details : CAS Number : 124-06-1 Weight : 0.02500kg

Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10403365

  • £11.40 / 25g


Estimated Shipment
21-05-2018
Add to basket

Specifications

Chemical Identifiers

Ethyl myristate
98+%
CH3(CH2)12COOCH2CH3
MFCD00008984
≥98%
C16H32O2
256.43
Ethyl tetradecanoate

Specifications

Ethyl myristate
295.0°C
11.0° to 12.0°C
124-06-1
Authentic
0.861
≥98%
C16H32O2
256.43
15, 6419
>110°C
Clear colorless
25g
0.8610g/mL
Glass bottle
Liquid, Solidifying In The Cold
98+%
CH3(CH2)12COOCH2CH3
MFCD00008984
Ethyl tetradecanoate
Videos
SDS