missing translation for 'onlineSavingsMsg'
Learn More

Ethyl myristate, 98+%

Product Code. 10187183 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10187183 500 g Glass bottle 500g
10403365 25 g Glass bottle 25g
10298943 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10187183 Supplier Thermo Scientific Acros Supplier No. 385545000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 124-06-1 | C16H32O2 | 256.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-06-1
Molecular Formula C16H32O2
Molecular Weight (g/mol) 256.43
MDL Number MFCD00008984
InChI Key MMKRHZKQPFCLLS-UHFFFAOYSA-N
Synonym ethyl myristate, tetradecanoic acid, ethyl ester, myristic acid ethyl ester, myristic acid, ethyl ester, ethylmyristate, ethyl n-tetradecanoate, ethyl myristate natural, fema no. 2445, tetradecanoic acid ethyl ester, ethyl ester tetradecanoic acid
PubChem CID 31283
ChEBI CHEBI:84849
IUPAC Name ethyl tetradecanoate
SMILES CCCCCCCCCCCCCC(=O)OCC

Specifications

Melting Point 11.0°C to 12.0°C
Color Colorless
Density 0.8610g/mL
Boiling Point 295.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)12COOCH2CH3
Quantity 500 g
Merck Index 15, 6419
Specific Gravity 0.861
Solubility Information Solubility in water: insoluble
Formula Weight 256.43
Percent Purity 98+%
Physical Form Liquid, Solidifying In The Cold
Chemical Name or Material Ethyl myristate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.