missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Mycophenolate, TRC

Product Code. 30383964
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30383964 500 mg 500mg
30368972 100 mg -
30371976 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30383964 Supplier Toronto Research Chemicals Supplier No. TRCE923345500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32483-51-5
Molecular Formula C19 H24 O6
Molecular Weight (g/mol) 348.39
Synonym (4E)-6-(1,3-Dihydro-4-hydroxy-6-methoxy-7-methyl-3-oxo-5-isobenzofuranyl)-4-methyl-4-hexenoic acid ethyl ester, 4-Hexenoic acid, 6-(1,3-dihydro-4-hydroxy-6-methoxy-7-methyl-3-oxo-5-isobenzofuranyl)-4-methyl-, ethyl ester, (E)-, Ethyl mycophenolate
IUPAC Name ethyl (E)-6-(4-hydroxy-6-methoxy-7-methyl-3-oxo-1H-2-benzofuran-5-yl)-4-methylhex-4-enoate
SMILES CCOC(=O)CC\C(=C\Cc1c(O)c2C(=O)OCc2c(C)c1OC)\C

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C19H24O6/c1-5-24-15(20)9-7-11(2)6-8-13-17(21)16-14(10-25-19(16)22)12(3)18(13)23-4/h6,21H,5,7-10H2,1-4H3/b11-6+
Formula Weight 348.1573
Chemical Name or Material Ethyl Mycophenolate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.