missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Mycophenolate, Mikromol™
Description
Ethyl Mycophenolate, Mikromol
Specifications
Specifications
| Chemical Name or Material | Ethyl Mycophenolate |
| CAS | 32483-51-5 |
| Analyte or Component Names | Ethyl Mycophenolate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C19 H24 O6 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Ethyl Mycophenolate, (4E)-6-(1,3-Dihydro-4-hydroxy-6-methoxy-7-methyl-3-oxo-5-isobenzofuranyl)-4-methyl-4-hexenoic acid ethyl ester, 4-Hexenoic acid, 6-(1,3-dihydro-4-hydroxy-6-methoxy-7-methyl-3-oxo-5-isobenzofuranyl)-4-methyl-, ethyl ester, (E)-, Ethyl mycophenolate |
| InChI Formula | InChI=1S/C19H24O6/c1-5-24-15(20)9-7-11(2)6-8-13-17(21)16-14(10-25-19(16)22)12(3)18(13)23-4/h6,21H,5,7-10H2,1-4H3/b11-6+ |
| Solution Type | Neat |
| SMILES | CCOC(=O)CC\C(=C\Cc1c(O)c2C(=O)OCc2c(C)c1OC)\C |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?