missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Mycophenolate, Mikromol™

Product Code. 18070980
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18070980 25 mg 25mg
18043670 100 mg 100mg
30462276 4 x 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18070980 Supplier Mikromol Supplier No. MM1001.090025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Ethyl Mycophenolate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl Mycophenolate
CAS 32483-51-5
Analyte or Component Names Ethyl Mycophenolate
Shipping Condition Room Temperature
Molecular Formula C19 H24 O6
Certifications/Compliance ISO 17025
Synonym Ethyl Mycophenolate, (4E)-6-(1,3-Dihydro-4-hydroxy-6-methoxy-7-methyl-3-oxo-5-isobenzofuranyl)-4-methyl-4-hexenoic acid ethyl ester, 4-Hexenoic acid, 6-(1,3-dihydro-4-hydroxy-6-methoxy-7-methyl-3-oxo-5-isobenzofuranyl)-4-methyl-, ethyl ester, (E)-, Ethyl mycophenolate
InChI Formula InChI=1S/C19H24O6/c1-5-24-15(20)9-7-11(2)6-8-13-17(21)16-14(10-25-19(16)22)12(3)18(13)23-4/h6,21H,5,7-10H2,1-4H3/b11-6+
SMILES CCOC(=O)CC\C(=C\Cc1c(O)c2C(=O)OCc2c(C)c1OC)\C
IUPAC Name ethyl (E)-6-(4-hydroxy-6-methoxy-7-methyl-3-oxo-1H-2-benzofuran-5-yl)-4-methylhex-4-enoate
Molecular Weight (g/mol) 348.39
Quantity 25 mg
Formula Weight 348.1573 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.