missing translation for 'onlineSavingsMsg'
Learn More

Ethyl morpholinoacetate, 98%, Thermo Scientific™

Catalog Number 10088841
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
3235-82-3
C8H15NO3
173.21
MFCD00023375
SITMDWHJQROIPF-UHFFFAOYSA-N
ethyl 2-morpholinoacetate, 4-morpholineacetic acid, ethyl ester, ethyl morpholinoacetate, ethyl 2-morpholin-4-yl acetate, morpholin-4-yl-acetic acid ethyl ester, ethyl 4-morpholineacetate, ethyl-2-morpholinoacetate, ethyl 4-morpholinylacetate, ethyl morpholin-4-ylacetate, morpholinoacetic acid ethyl ester
2734105
ethyl 2-morpholin-4-ylacetate
CCOC(=O)CN1CCOCC1
This item is not returnable. View return policy
3235-82-3
C8H15NO3
173.21
MFCD00023375
SITMDWHJQROIPF-UHFFFAOYSA-N
ethyl 2-morpholinoacetate, 4-morpholineacetic acid, ethyl ester, ethyl morpholinoacetate, ethyl 2-morpholin-4-yl acetate, morpholin-4-yl-acetic acid ethyl ester, ethyl 4-morpholineacetate, ethyl-2-morpholinoacetate, ethyl 4-morpholinylacetate, ethyl morpholin-4-ylacetate, morpholinoacetic acid ethyl ester
2734105
ethyl 2-morpholin-4-ylacetate
CCOC(=O)CN1CCOCC1
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4535 to 1.4555
Quantity 5 g
Formula Weight 173.21
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethyl morpholinoacetate, 98%
compliance-icons
Product Identifier
  • Ethyl morpholinoacetate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'