missing translation for 'onlineSavingsMsg'
Learn More

Ethyl morpholinoacetate, 98%, Thermo Scientific™

Product Code. p-130932 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10088841 5 g Glass bottle 5g
10742633 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10088841 Supplier Thermo Scientific Acros Supplier No. 350910050
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 3235-82-3
Molecular Formula C8H15NO3
Molecular Weight (g/mol) 173.21
MDL Number MFCD00023375
InChI Key SITMDWHJQROIPF-UHFFFAOYSA-N
Synonym ethyl 2-morpholinoacetate, 4-morpholineacetic acid, ethyl ester, ethyl morpholinoacetate, ethyl 2-morpholin-4-yl acetate, morpholin-4-yl-acetic acid ethyl ester, ethyl 4-morpholineacetate, ethyl-2-morpholinoacetate, ethyl 4-morpholinylacetate, ethyl morpholin-4-ylacetate, morpholinoacetic acid ethyl ester
PubChem CID 2734105
IUPAC Name ethyl 2-morpholin-4-ylacetate
SMILES CCOC(=O)CN1CCOCC1

Specifications

Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4535 to 1.4555
Quantity 5 g
Formula Weight 173.21
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethyl morpholinoacetate, 98%
compliance-icons
Product Identifier
  • Ethyl morpholinoacetate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.