missing translation for 'onlineSavingsMsg'
Learn More

Ethyl methylcarbamate, 99%

Product Code. 10742402 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10742402 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10742402 Supplier Thermo Scientific Acros Supplier No. 410260050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 105-40-8 | C4H9NO2 | 103.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 105-40-8
Boiling Point 170.0°C
Assay Percent Range 98.5% min. (GC)
Molecular Formula C4H9NO2
Packaging Glass Bottle
Linear Formula CH3NHCOOC2H5
Refractive Index 1.4180 to 1.4200
MDL Number MFCD00041924
Synonym ethyl methylcarbamate, n-methylurethane, n-methylurethan, ethyl-n-methylcarbamate, methylcarbamic acid, ethyl ester, n-methyl urethan, carbamic acid, methyl-, ethyl ester, methylurethane van, unii-1w34gcf5cs, ccris 2510
Solubility Information Solubility in water: 6900g/L (16°C). Other solubilities: soluble in alcohol
InChI Key SURZCVYFPAXNGN-UHFFFAOYSA-N
Density 1.0100g/mL
SMILES CCOC(=O)NC
IUPAC Name ethyl N-methylcarbamate
Molecular Weight (g/mol) 103.12
Quantity 5 g
Flash Point 61°C
PubChem CID 7752
Formula Weight 103.12
Percent Purity 99%
Specific Gravity 1.01
Chemical Name or Material Ethyl methylcarbamate, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl methylcarbamate
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.