missing translation for 'onlineSavingsMsg'
Learn More

Ethyl methacrylate, 98+%, stab. with 10-120ppm, 4-methoxyphenol

Product Code. 11428137
Change view
Click to view available options
Quantity:
250 mL
1000 mL
Unit Size:
1000mL
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11428137 250 mL 250mL
11418137 1000 mL 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11428137 Supplier Thermo Scientific Alfa Aesar Supplier No. A17518.AK
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 97-63-2 | C6H10O2 | 114.144 g/mol

Ethyl methacrylate is used to prepare copolymers of EMA on waxy maize starch and hydroxypropylstarch, potential application of these copolymers are as excipients for compressed non-disintegrating matrix tablets. It is also used to study reactions of n-butyl acrylate and EMA with ozone in the gas phase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl methacrylate is used to prepare copolymers of EMA on waxy maize starch and hydroxypropylstarch, potential application of these copolymers are as excipients for compressed non-disintegrating matrix tablets. It is also used to study reactions of n-butyl acrylate and EMA with ozone in the gas phase.

Solubility
Soluble in water (4.69g/L), ether, and alcohols.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-63-2
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.144
MDL Number MFCD00009161
InChI Key SUPCQIBBMFXVTL-UHFFFAOYSA-N
Synonym ethyl methacrylate, ethyl 2-methylacrylate, 2-propenoic acid, 2-methyl-, ethyl ester, methacrylic acid ethyl ester, ethyl 2-methyl-2-propenoate, ethyl 2-methacrylate, rhoplex ac-33, methacrylic acid, ethyl ester, rcra waste number u118, 2-methylacrylic acid, ethyl ester
PubChem CID 7343
IUPAC Name ethyl 2-methylprop-2-enoate
SMILES CCOC(=O)C(=C)C

Specifications

Melting Point -60°C
Density 0.917
Boiling Point 118°C to 119°C
Flash Point 18°C (64°F)
Odor Pungent
Refractive Index 1.414
Quantity 250 mL
UN Number UN2277
Beilstein 471201
Solubility Information Soluble in water (4.69g/L),ether,and alcohols.
Formula Weight 114.15
Percent Purity ≥98%
Chemical Name or Material Ethyl methacrylate, stabilized with 100ppm 4-methoxyphenol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethyl methacrylate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Ethyl methacrylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.