missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-Ethyl mandelate, 97%

Product Code. 11408007
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11408007

Brand: Thermo Scientific Alfa Aesar A17412.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 774-40-3 | C10H12O3 | 180.203 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 774-40-3
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00004494
InChI Key SAXHIDRUJXPDOD-UHFFFAOYSA-N
Synonym ethyl mandelate, ethyl phenylglycolate, mandelic acid, ethyl ester, dl-mandelic acid ethyl ester, mandelic acid ethyl ester, ethyl dl-mandelate, mandelsaeureaethylester, ethyl hydroxy phenyl acetate, mandelsaeureaethylester german, +-ethyl mandelate
PubChem CID 13050
ChEBI CHEBI:38750
IUPAC Name ethyl 2-hydroxy-2-phenylacetate
SMILES CCOC(=O)C(C1=CC=CC=C1)O

Specifications

Density 1.115
Boiling Point 253°C to 255°C
Flash Point >110°C (230°F)
Quantity 25 g
Formula Weight 180.2
Percent Purity 97%
Chemical Name or Material (+/-)-Ethyl mandelate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.