missing translation for 'onlineSavingsMsg'
Learn More

Ethyl maltol, 98%

Product Code. p-7077351 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 15254258

Brand: Thermo Scientific Acros 450800250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4940-11-8 | C7H8O3 | 140.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11-8-4940
Molecular Formula C7H8O3
Molecular Weight (g/mol) 140.14

Specifications

Melting Point 89.0°C to 93.0°C
Boiling Point 289.0°C to 290.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 25 g
Merck Index 15,3879
Synonym ethyl maltol, 2-ethyl-3-hydroxy-4h-pyran-4-one, 2-ethyl-3-hydroxy-4-pyrone, veltol plus, 3-hydroxy-2-ethyl-4-pyrone, ethylmaltol, 2-ethylpyromeconic acid, 4h-pyran-4-one, 2-ethyl-3-hydroxy, 3-hydroxy-2-ethyl-1,4-pyrone, 2-ethyl pyromeconic acid
InChI Key YIKYNHJUKRTCJL-UHFFFAOYSA-N
SMILES CCC1=C(C(=O)C=CO1)O
IUPAC Name 2-ethyl-3-hydroxypyran-4-one
PubChem CID 21059
Formula Weight 140.14
Percent Purity 98%
Chemical Name or Material Ethyl maltol
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl maltol, 98%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.