missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Malonyl Chloride, TRC

Product Code. 30387017
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30387017 250 mg
30382420 1 g
30388140 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30387017 Supplier Toronto Research Chemicals Supplier No. TRCE922050250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36239-09-5
Molecular Formula C5H7ClO3
Molecular Weight (g/mol) 150.56
Synonym (Chlorocarbonyl)acetic Acid Ethyl Ester, (Chloroformyl)acetic Acid Ethyl Ester, (Ethoxycarbonyl)acetyl Chloride, 2-(Ethoxycarbonyl)acetyl Chloride, 3-Chloro-3-oxopropanoic Acid Ethyl Ester, 3-Ethoxy-3-oxopropanoyl Chloride, Carbethoxyacetyl Chloride, Carbethoxyethanoyl Chloride, Carboethoxyacetyl Chloride, Ethoxymalonyl Chloride, Ethyl (Chlorocarbonyl)acetate, Ethyl (Chloroformyl)acetate, Ethyl 3-Chloro-3-oxopropanoate, Ethyl 3-Chloro-3-oxopropionate, Ethyl 3-Oxo-3-chloropropanoate, Ethyl Malonoyl Chloride, Malonic Acid Chloride Ethyl Ester, Malonic acid chloride monoethyl ester
IUPAC Name ethyl 3-chloro-3-oxopropanoate
SMILES ClC(CC(OCC)=O)=O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C5H7ClO3/c1-2-9-5(8)3-4(6)7/h2-3H2,1H3
Formula Weight 150.01
Chemical Name or Material Ethyl Malonyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.