missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Ethyl LipotF
GREENER_CHOICE

Product Code. 18778426
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18778426 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18778426 Supplier Tocris Bioscience™ Supplier No. 5950/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective FTO inhibitor

Ethyl LipotF is a selective FTO inhibitor (IC50 = 0.81 μM). Exhibits >30-fold selectivity for FTO over ALKBH2, 3 and 5 and >100-fold selectivity over PHD2 and JMJD2A. Increases levels of modified N6-methyladenosine (m6A) in HeLa cells in a concentration-dependent manner.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl (E)-4-(2-(4-benzylnicotinoyl)hydrazinyl)-4-oxobut-2-enoate
CAS 1808089-27-1
Quantity 10 mg
Target Other Oxidase/Oxygenase Compounds
Molecular Formula C19H19N3O4
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.