missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Levulinate, TRC
Click to view available options
Quantity:
500 g
Unit Size:
500g
Specifications
Specifications
| Chemical Name or Material | Ethyl Levulinate |
| CAS | 539-88-8 |
| Molecular Formula | C7H12O3 |
| Quantity | 500 g |
| Synonym | 4-Oxopentanoic Acid Ethyl Ester, Levulinic Acid Ethyl Ester, 4-Oxopentanoic Acid Ethyl Ester, Ethyl 3-Acetylpropionate, Ethyl 4-Ketovalerate, Ethyl 4-Oxopentanoate, Ethyl 4-Oxovalerate, Ethyl Laevulinate, NSC 24876, NSC 8863 |
| InChI Formula | InChI=1S/C7H12O3/c1-3-10-7(9)5-4-6(2)8/h3-5H2,1-2H3 |
| SMILES | CCOC(=O)CCC(=O)C |
| IUPAC Name | ethyl 4-oxopentanoate |
| Molecular Weight (g/mol) | 144.17 |
| Formula Weight | 144.0786 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction