missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Levulinate, TRC

Product Code. 30372712
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
This item is not returnable. View return policy

Product Code. 30372712

Brand: Toronto Research Chemicals TRCE9219651KG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl Levulinate
CAS 539-88-8
Molecular Formula C7H12O3
Quantity 1 kg
Synonym 4-Oxopentanoic Acid Ethyl Ester, Levulinic Acid Ethyl Ester, 4-Oxopentanoic Acid Ethyl Ester, Ethyl 3-Acetylpropionate, Ethyl 4-Ketovalerate, Ethyl 4-Oxopentanoate, Ethyl 4-Oxovalerate, Ethyl Laevulinate, NSC 24876, NSC 8863
InChI Formula InChI=1S/C7H12O3/c1-3-10-7(9)5-4-6(2)8/h3-5H2,1-2H3
SMILES CCOC(=O)CCC(=O)C
IUPAC Name ethyl 4-oxopentanoate
Molecular Weight (g/mol) 144.17
Formula Weight 144.0786
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.