missing translation for 'onlineSavingsMsg'
Learn More

Ethyl levulinate, 98%

Product Code. 11457227
Change view
Click to view available options
Quantity:
50 g
100 g
500 g
Unit Size:
100g
500g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11457227 50 g 50g
11488930 100 g 100g
11498930 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11457227 Supplier Thermo Scientific Alfa Aesar Supplier No. A15001.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 539-88-8 | C7H12O3 | 144.17 g/mol

Ethyl levulinate is used to develop a preparative HPLC method to preserve wine aromas and isolates fruity characteristics of red wine aroma extracts

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl levulinate is used to develop a preparative HPLC method to preserve wine aromas and isolates fruity characteristics of red wine aroma extracts

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 539-88-8
Molecular Formula C7H12O3
Molecular Weight (g/mol) 144.17
MDL Number MFCD00009209
InChI Key GMEONFUTDYJSNV-UHFFFAOYSA-N
Synonym ethyl levulinate, ethyl levulate, ethyl laevulinate, levulinic acid, ethyl ester, ethyl 4-ketovalerate, ethyl 3-acetylpropionate, ethyl 4-oxovalerate, pentanoic acid, 4-oxo-, ethyl ester, ethyl ketovalerate, ethyl acetylpropanoate
PubChem CID 10883
IUPAC Name ethyl 4-oxopentanoate
SMILES CCOC(=O)CCC(C)=O

Specifications

Density 1.014
Boiling Point 203°C to 205°C
Flash Point 94°C (201°F)
Refractive Index 1.422
Quantity 50 g
Beilstein 507641
Merck Index 14,3819
Solubility Information Soluble in water.
Formula Weight 144.17
Percent Purity 98%
Chemical Name or Material Ethyl levulinate
Show More Show Less
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.