missing translation for 'onlineSavingsMsg'
Learn More

Ethyl levulinate, 98%

Product Code. 10581781 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10581781 500 g Glass bottle 500g
10163390 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10581781 Supplier Thermo Scientific Acros Supplier No. 153625000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 539-88-8 | C7H12O3 | 144.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 539-88-8
Molecular Formula C7H12O3
Molecular Weight (g/mol) 144.17
MDL Number MFCD00009209
InChI Key GMEONFUTDYJSNV-UHFFFAOYSA-N
Synonym ethyl levulinate, ethyl levulate, ethyl laevulinate, levulinic acid, ethyl ester, ethyl 4-ketovalerate, ethyl 3-acetylpropionate, ethyl 4-oxovalerate, pentanoic acid, 4-oxo-, ethyl ester, ethyl ketovalerate, ethyl acetylpropanoate
PubChem CID 10883
IUPAC Name ethyl 4-oxopentanoate
SMILES CCOC(=O)CCC(C)=O

Specifications

Acidity 5mg KOH/g max.
Color Yellow
Density 1.0120g/mL
Boiling Point 206°C
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3COCH2CH2CO2C2H5
Refractive Index 1.4210 to 1.423
Quantity 500 g
Beilstein 03, 675
Merck Index 15, 3874
Specific Gravity 1.012
Solubility Information Solubility in water: 40g/L(20°C). Other solubilities: miscible with alcohol
Formula Weight 144.17
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethyl levulinate, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.