missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Lauroyl Arginate-d23 Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | Ethyl Lauroyl Arginate-d23 Hydrochloride |
| Molecular Formula | C20 D23 H17 N4 O3 . H Cl |
| Quantity | 10 mg |
| Synonym | N2-(1-Oxododecyl)-L-arginine Ethyl Ester-d23 Hydrochloride, N2-(1-Oxododecyl)-L-arginine Ethyl Ester-d23 Monohydrochlorid, Aminat G-d23, CytoGuard LA-d23, Ethyl-Nalpha-lauroyl-L-arginate-d23 Hydrochloride, Mirenat CF-d23, Mirenat-N-d23 |
| InChI Formula | InChI=1S/C20H40N4O3.ClH/c1-3-5-6-7-8-9-10-11-12-15-18(25)24-17(19(26)27-4-2)14-13-16-23-20(21)22;/h17H,3-16H2,1-2H3,(H,24,25)(H4,21,22,23);1H/t17-;/m0./s1/i1D3,3D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,15D2; |
| SMILES | Cl.[2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C(=O)N[C@@H](CCCNC(=N)N)C(=O)OCC |
| IUPAC Name | ethyl (2S)-5-carbamimidamido-2-(2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-tricosadeuteriododecanoylamino)pentanoate;hydrochloride |
| Molecular Weight (g/mol) | 444.159 |
| Formula Weight | 443.431 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?