missing translation for 'onlineSavingsMsg'
Learn More

(-)-Ethyl L-Lactate, TRC

Product Code. 30134441
Change view
Click to view available options
Quantity:
1 g
100 g
Unit Size:
100g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134441 100 g 100g
17973195 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134441 Supplier Toronto Research Chemicals Supplier No. TRCE921970100G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 687-47-8
Molecular Formula C5 H10 O3
Molecular Weight (g/mol) 118.131
Synonym Lactic acid, ethyl ester, (-)- (8CI), Propanoic acid, 2-hydroxy-, ethyl ester, (S)-, (-)-Ethyl 2-hydroxypropanoate, (-)-Ethyl 2-hydroxypropionate, (-)-Ethyl lactate, (2S)-2-Hydroxypropanoic acid ethyl ester, (2S)-2-Hydroxypropionic acid ethyl ester, (L)-(-)-Ethyl lactate, (S)-(-)-Ethyl lactate, (S)-(-)-Lactic acid ethyl ester, (S)-2-Hydroxypropanoic acid ethyl ester, (S)-Lactic acid ethyl ester, Ethyl (-)-(S)-lactate, Ethyl (2S)-(-)-2-hydroxypropanoate, Ethyl (2S)-2-hydroxypropanoate, Ethyl (S)-(-)-2-hydroxypropionate, Ethyl (S)-2-hydroxypropanoate, Ethyl (S)-2-hydroxypropionate, Ethyl (S)-lactate, Ethyl L-(-)-lactate, Ethyl L-lactate, L-Ethyl lactate
IUPAC Name ethyl (2S)-2-hydroxypropanoate
SMILES CCOC(=O)[C@H](C)O

Specifications

Quantity 100 g
InChI Formula InChI=1S/C5H10O3/c1-3-8-5(7)4(2)6/h4,6H,3H2,1-2H3/t4-/m0/s1
Formula Weight 118.063
Chemical Name or Material (S)-Lactic acid-ethyl ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.