missing translation for 'onlineSavingsMsg'
Learn More

Ethyl L-lactate, 99%

Product Code. p-7081087
Click to view available options
Quantity:
10,000 g
2500 g
500 g
Unit Size:
10000g
2500g
500g
This item is not returnable. View return policy

Product Code. 11474263

Brand: Thermo Scientific Alfa Aesar A10900.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 687-47-8 | C5H10O3 | 118.132 g/mol

Ethyl L-lactate is used in the preparation of food additives, aryl aldimines, fragrances and in pharmaceutical preparations. It can be considered as a green solvent in chemical industries due to its biodegradable nature. As a solvent, it used in the production of nitrocellulose, cellulose acetate and cellulose ethers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl L-lactate is used in the preparation of food additives, aryl aldimines, fragrances and in pharmaceutical preparations. It can be considered as a green solvent in chemical industries due to its biodegradable nature. As a solvent, it used in the production of nitrocellulose, cellulose acetate and cellulose ethers.

Solubility
Miscible with water and chloroform.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 687-47-8
Molecular Formula C5H10O3
Molecular Weight (g/mol) 118.132
MDL Number MFCD00065359
InChI Key LZCLXQDLBQLTDK-BYPYZUCNSA-N
Synonym ethyl l-lactate, ethyl 2s-2-hydroxypropanoate, l-ethyl lactate, l---ethyl lactate, ethyl l--lactate, ethyl s-2-hydroxypropionate, ethyl l---lactate, s-ethyl lactate, propanoic acid, 2-hydroxy-, ethyl ester, 2s
PubChem CID 92831
ChEBI CHEBI:78322
IUPAC Name ethyl (2S)-2-hydroxypropanoate
SMILES CCOC(=O)C(C)O

Specifications

Melting Point °C to 25°C
Density 1.036 g/mL
Boiling Point 152°C to 155°C
Flash Point 46°C (114°F)
Assay Percent Range 99%
Refractive Index 1.413
Quantity 500 g
UN Number UN1192
Beilstein 1720839
Merck Index 14,3817
Solubility Information Miscible with water and chloroform.
Formula Weight 118.13
Percent Purity 99%
Chemical Name or Material Ethyl L-lactate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ethyl L-lactate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Ethyl (S)-2-hydroxypropionate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.