Learn More
Ethyl L-lactate, 99%
Description
Ethyl L-lactate is used in the preparation of food additives, aryl aldimines, fragrances and in pharmaceutical preparations. It can be considered as a green solvent in chemical industries due to its biodegradable nature. As a solvent, it used in the production of nitrocellulose, cellulose acetate and cellulose ethers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ethyl L-lactate is used in the preparation of food additives, aryl aldimines, fragrances and in pharmaceutical preparations. It can be considered as a green solvent in chemical industries due to its biodegradable nature. As a solvent, it used in the production of nitrocellulose, cellulose acetate and cellulose ethers.
Solubility
Miscible with water and chloroform.
Notes
Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | -25°C |
| Density | 1.036 |
| Boiling Point | 152°C to 155°C |
| Flash Point | 46°C (114°F) |
| Assay Percent Range | 99% |
| Refractive Index | 1.413 |
| Quantity | 2500 g |
| UN Number | UN1192 |
| Beilstein | 1720839 |
| Merck Index | 14,3817 |
| Show More |
Safety and Handling
- Ethyl L-lactate
- Danger
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Specific target organ toxicity Category 3
- H226-Flammable liquid and vapour.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contain: Ethyl (S)-2-hydroxypropionate
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.