missing translation for 'onlineSavingsMsg'
Learn More

Ethyl L(-)-lactate, 97%

Product Code. 10142290 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
Unit Size:
1 L
10 L
2.5 L
250mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10142290 1 L Glass bottle 1 L
10561774 250 mL Glass bottle 250mL
10042353 2.5 L Glass bottle 2.5 L
13418328 10 L Plastic drum 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10142290 Supplier Thermo Scientific Acros Supplier No. 118660010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 687-47-8 | C5H10O3 | 118.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 687-47-8
Molecular Formula C5H10O3
Molecular Weight (g/mol) 118.13
MDL Number MFCD00004518
InChI Key LZCLXQDLBQLTDK-BYPYZUCNSA-N
Synonym ethyl l-lactate, ethyl 2s-2-hydroxypropanoate, l-ethyl lactate, l---ethyl lactate, ethyl l--lactate, ethyl s-2-hydroxypropionate, ethyl l---lactate, s-ethyl lactate, propanoic acid, 2-hydroxy-, ethyl ester, 2s
PubChem CID 92831
ChEBI CHEBI:78322
IUPAC Name ethyl (2S)-2-hydroxypropanoate
SMILES CCOC(=O)C(C)O

Specifications

Acidity 0.1% max. (as lactic acid)
Melting Point -26.0°C
Density 1.0340g/mL
Boiling Point 154.0°C
Flash Point 46°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH(OH)CO2C2H5
Refractive Index 1.4100 to 1.4160
Quantity 1 L
Beilstein 03,264
Fieser 17,135
Merck Index 14,3817
Specific Gravity 1.034
Specific Rotation Condition − 10.00 (20.00°C neat)
Specific Rotation − 10.00
Solubility Information Solubility in water: soluble. Other solubilities: miscible with alcohols,ketones and esters
Formula Weight 118.13
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Ethyl L(-)-lactate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ethyl L(-)-lactate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.