missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Isovalerate(Isovaleric Acid Ethyl Ester), TRC

Product Code. 30006322
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30006322 5 g 5g
30006188 25 g 25g
30004387 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30006322 Supplier Toronto Research Chemicals Supplier No. TRCI9175755G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-64-5
Molecular Formula C7 H14 O2
Molecular Weight (g/mol) 130.18
Synonym Isovaleric Acid-Ethyl Ester, Butanoic acid, 3-methyl-, ethyl ester, Isovaleric acid, ethyl ester (6CI,7CI,8CI), 3-Methylbutanoic acid ethyl ester, 3-Methylbutyric acid ethyl ester, Ethyl 3-methylbutanoate, Ethyl 3-methylbutyrate, Ethyl isopentanoate, Ethyl isovalerate, Isopentanoic acid ethyl ester, NSC 8869
IUPAC Name ethyl 3-methylbutanoate
SMILES CCOC(=O)CC(C)C

Specifications

Quantity 5 g
InChI Formula InChI=1S/C7H14O2/c1-4-9-7(8)5-6(2)3/h6H,4-5H2,1-3H3
Formula Weight 130.0994
Chemical Name or Material Ethylisovalerate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.