missing translation for 'onlineSavingsMsg'
Learn More

Ethyl isovalerate, 98%

Product Code. 11434033
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Unit Size:
100mL
2500mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11434033 500 mL 500mL
11424033 100 mL 100mL
11444033 2500 mL 2500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11434033 Supplier Thermo Scientific Alfa Aesar Supplier No. A10292.AP
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 108-64-5 | C7H14O2 | 130.187 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-64-5
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.187
MDL Number MFCD00009203
InChI Key PPXUHEORWJQRHJ-UHFFFAOYSA-N
Synonym ethyl isovalerate, ethyl 3-methylbutyrate, ethylisovalerate, ethyl isopentanoate, isovaleric acid, ethyl ester, butanoic acid, 3-methyl-, ethyl ester, ethyl beta-methylbutyrate, ethyl isovalerianate, 3-methylbutyric acid ethyl ester, 3-methylbutanoic acid ethyl ester
PubChem CID 7945
ChEBI CHEBI:31571
IUPAC Name ethyl 3-methylbutanoate
SMILES CCOC(=O)CC(C)C

Specifications

Melting Point -99°C
Density 0.866
Boiling Point 131°C to 133°C
Flash Point 26°C (78°F)
Odor Fruit-like
Refractive Index 1.396
Quantity 500 mL
UN Number UN3272
Beilstein 1744677
Merck Index 14,3816
Formula Weight 130.19
Percent Purity 98%
Chemical Name or Material Ethyl isovalerate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.