missing translation for 'onlineSavingsMsg'
Learn More

Ethyl isonitrosocyanoacetate, 97%

Product Code. 11480593
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11480593 100 g 100g
11460593 5 g 5g
11470593 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11480593 Supplier Thermo Scientific Alfa Aesar Supplier No. A12691.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 3849-21-6 | C5H6N2O3 | 142.114 g/mol

This product is a non-explosive alternative to HOBt or HOAt, and allows high peptide coupling rates at low racemization when applied in combination with carbodiimides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3849-21-6
Molecular Formula C5H6N2O3
Molecular Weight (g/mol) 142.114
MDL Number MFCD00000625
InChI Key LCFXLZAXGXOXAP-QPJJXVBHSA-N
Synonym ethyl cyanoglyoxylate-2-oxime, ethyl isonitrosocyanoacetate, ethyl cyano hydroxyimino acetate, ethyl hydroxyimino cyanoacetate, ethyl 2-cyano-2-hydroxyimino acetate, ethyl cyano hydroxyimino formate, ethyl oximinocyanoacetate, oxyma, e-ethyl 2-cyano-2-hydroxyimino acetate, acetic acid, cyano hydroxyimino-, ethyl ester
PubChem CID 6400537
IUPAC Name ethyl (2E)-2-cyano-2-hydroxyiminoacetate
SMILES CCOC(=O)C(=NO)C#N

Specifications

Melting Point 127°C to 132°C
Quantity 100 g
Formula Weight 142.11
Percent Purity 97%
Chemical Name or Material Ethyl isonitrosocyanoacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.