missing translation for 'onlineSavingsMsg'
Learn More

Ethyl isodehydroacetate, 95%, Thermo Scientific™

Product Code. 10301173 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10301173 5 g Glass bottle 5g
10421543 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10301173 Supplier Thermo Scientific Acros Supplier No. 373900050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3385-34-0
Molecular Formula C10H12O4
Molecular Weight (g/mol) 196.20
MDL Number MFCD00006643
InChI Key FBPWNVQUVXSXKS-UHFFFAOYSA-N
Synonym ethyl isodehydroacetate, ethyl 4,6-dimethyl-2-oxo-2h-pyran-5-carboxylate, ethyl isodehydracetate, 5-carbethoxy-4,6-dimethyl-2-pyrone, 2h-pyran-5-carboxylic acid, 4,6-dimethyl-2-oxo-, ethyl ester, ethyl 4,6-dimethyl-5-coumalate, 2,4-dimethyl-6-oxo-6h-pyran-3-carboxylic acid ethyl ester, ethylisodehydroacetate, ethyl 4,6-dimethyl-2-oxopyran-5-carboxylate, acmc-20am57
PubChem CID 76918
IUPAC Name ethyl 2,4-dimethyl-6-oxopyran-3-carboxylate
SMILES CCOC(=O)C1=C(C)OC(=O)C=C1C

Specifications

Melting Point 17°C to 18°C
Density 1.1670g/mL
Boiling Point 290°C to 295°C
Flash Point >110°C
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.5135 to 1.5165
Quantity 5 g
Beilstein 18,41
Specific Gravity 1.167
Solubility Information Solubility in water: insoluble
Formula Weight 196.2
Percent Purity 95%
Chemical Name or Material Ethyl isodehydroacetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.