missing translation for 'onlineSavingsMsg'
Learn More

Ethyl isocyanatoacetate, 98%

Product Code. 10264724 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10264724 5 g Glass bottle 5g
10577254 25 g Glass bottle 25g
10192764 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10264724 Supplier Thermo Scientific Acros Supplier No. 410150050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2949-22-6 | C5H7NO3 | 129.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2949-22-6
Molecular Formula C5H7NO3
Molecular Weight (g/mol) 129.12
MDL Number MFCD00002040
InChI Key DUVOZUPPHBRJJO-UHFFFAOYSA-N
Synonym ethyl isocyanatoacetate, isocyanatoacetic acid ethyl ester, acetic acid, isocyanato-, ethyl ester, carbethoxymethyl isocyanate, isocyanatoethyl acetate, ethoxycarbonylmethyl isocyanate, ethylisocyanatoacetate, ethyl glycolate isocyanate, ethyl n-oxomethylene glycinate, acetic acid,2-isocyanato-, ethyl ester
PubChem CID 76283
IUPAC Name ethyl 2-isocyanatoacetate
SMILES CCOC(=O)CN=C=O

Specifications

Color Colorless
Density 1.1510g/mL
Boiling Point 67°C to 68°C (11.0 mmHg)
Flash Point 72°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula OCNCH2COOC2H5
Refractive Index 1.4210 to 1.4230 (20°C, 589nm)
Quantity 5 g
Specific Gravity 1.151
Solubility Information Solubility in water: hydrolyses
Formula Weight 129.11
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethyl isocyanatoacetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.