missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Isobutyrylacetate, TRC

Product Code. 30378569
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30378569 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30378569 Supplier Toronto Research Chemicals Supplier No. TRCE92130050G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl Isobutyrylacetate
CAS 7152-15-0
Molecular Formula C8H14O3
Quantity 50 g
Synonym 3-Oxo-4-methylpentanoic Acid Ethyl Ester, 4-Methyl-3-oxo-pentanoic Acid Ethyl Ester, 4-Methyl-3-oxovaleric Acid Ethyl Ester, Ethyl 2-isobutyrylacetate, Ethyl 3-Isopropyl-3-oxopropanoate, Ethyl 3-oxo-4-methylpentanoate, Ethyl 3-Oxo-4-methylvalerate, Ethyl 4-Methyl-3-oxopentanoate, Ethyl 4-Methyl-3-oxovalerate, Ethyl Isobutyrylacetate, Isobutyrylacetic Acid Ethyl Ester, NSC 62029, gamma,gamma-Dimethylacetoacetic Acid Ethyl Ester, Ethyl 4-Methyl-3-oxopentanoate
InChI Formula InChI=1S/C8H14O3/c1-4-11-8(10)5-7(9)6(2)3/h6H,4-5H2,1-3H3
SMILES CCOC(=O)CC(=O)C(C)C
IUPAC Name ethyl 4-methyl-3-oxopentanoate
Molecular Weight (g/mol) 158.19
Formula Weight 158.0943
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.