missing translation for 'onlineSavingsMsg'
Learn More

Ethyl isobutyrylacetate, 95%, Thermo Scientific Chemicals

Catalog Number 10413612
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
7152-15-0
C8H14O3
158.2
MFCD00009198
XCLDSQRVMMXWMS-UHFFFAOYSA-N
ethyl isobutyrylacetate, 4-methyl-3-oxo-pentanoic acid ethyl ester, pentanoic acid, 4-methyl-3-oxo-, ethyl ester, ethyl isobutyroylacetate, valeric acid, 4-methyl-3-oxo-, ethyl ester, .gamma.,.gamma.-dimethylacetoacetic acid ethyl ester, beta-keto-gamma-methylvaleric acid ethyl ester, ethyl alpha-isobutyroylacetate, ethylisobutyrylacetate, ethyl iso-butyrylacetate
81583
ethyl 4-methyl-3-oxopentanoate
CCOC(=O)CC(=O)C(C)C
This item is not returnable. View return policy
7152-15-0
C8H14O3
158.2
MFCD00009198
XCLDSQRVMMXWMS-UHFFFAOYSA-N
ethyl isobutyrylacetate, 4-methyl-3-oxo-pentanoic acid ethyl ester, pentanoic acid, 4-methyl-3-oxo-, ethyl ester, ethyl isobutyroylacetate, valeric acid, 4-methyl-3-oxo-, ethyl ester, .gamma.,.gamma.-dimethylacetoacetic acid ethyl ester, beta-keto-gamma-methylvaleric acid ethyl ester, ethyl alpha-isobutyroylacetate, ethylisobutyrylacetate, ethyl iso-butyrylacetate
81583
ethyl 4-methyl-3-oxopentanoate
CCOC(=O)CC(=O)C(C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -9.0°C
Color Colorless to Yellow
Density 0.9810g/mL
Boiling Point 173.0°C
Flash Point 44°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (Mixture of tautomers) (GC)
Packaging Glass bottle
Linear Formula (CH3)2CHCOCH2CO2C2H5
Refractive Index 1.4255 to 1.4275
Quantity 10 g
Specific Gravity 0.981
Formula Weight 158.2
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Ethyl isobutyrylacetate
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl isobutyrylacetate
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'