missing translation for 'onlineSavingsMsg'
Learn More

Ethyl isobutyrate, 99%

Product Code. 10472261 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass bottle
Unit Size:
2.5 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10472261 500 mL Glass bottle 500mL
15169263 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10472261 Supplier Thermo Scientific Acros Supplier No. 310515000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 97-62-1 | C6H12O2 | 116.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-62-1
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.18
MDL Number MFCD00009165
InChI Key WDAXFOBOLVPGLV-UHFFFAOYSA-N
Synonym ethyl isobutyrate, ethyl isobutanoate, ethylisobutyrate, isobutyric acid ethyl ester, ethyl 2,2-dimethylacetate, propanoic acid, 2-methyl-, ethyl ester, isobutyric acid, ethyl ester, ethyl 2-methylpropionate, propionic acid, 2-methyl-, ethyl ester, ethyl isobutyrate natural
PubChem CID 7342
ChEBI CHEBI:87303
IUPAC Name ethyl 2-methylpropanoate
SMILES CCOC(=O)C(C)C

Specifications

Color Colorless
Density 0.8600g/mL
Boiling Point 112.0°C to 113.0°C
Flash Point 13°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (CH3)2CHCO2C2H5
Refractive Index 1.3860 to 1.3880
Quantity 500 mL
Merck Index 15,3869
Specific Gravity 0.86
Solubility Information Solubility in water: not miscible
Formula Weight 116.18
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Ethyl isobutyrate, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.