missing translation for 'onlineSavingsMsg'
Learn More

Ethyl iodoacetate, 98%, stabilized, Thermo Scientific Chemicals

Product Code. 10338120 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
50 mL
250 mL
Packaging:
Glass bottle
Unit Size:
10mL
250mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10338120 50 mL Glass bottle 50mL
10501861 10 mL Glass bottle 10mL
10656594 250 mL Glass bottle 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10338120 Supplier Thermo Scientific Acros Supplier No. 222190500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 623-48-3
Molecular Formula C4H7IO2
Molecular Weight (g/mol) 214.00
MDL Number MFCD00001081
InChI Key MFFXVVHUKRKXCI-UHFFFAOYSA-N
Synonym ethyl iodoacetate, acetic acid, iodo-, ethyl ester, ethyl monoiodoacetate, iodoacetic acid, ethyl ester, acetic acid, 2-iodo-, ethyl ester, ethylester kyseliny jodoctove czech, acetic acid, 2-iodo-,ethyl ester, ethylester kyseliny jodoctove, ethyliodoacetate, iodoacetic acid ethyl
PubChem CID 12183
IUPAC Name ethyl 2-iodoacetate
SMILES CCOC(=O)CI

Specifications

Color Orange to Yellow
Density 1.8000g/mL
Boiling Point 179°C to 180°C
Flash Point 76°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula ICH2CO2C2H5
Refractive Index 1.5035 to 1.5055
Quantity 50 mL
Beilstein 02,222
Specific Gravity 1.8
Solubility Information Solubility in water: immiscible
Stabilizer copper
Formula Weight 214
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethyl iodoacetate, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.