missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Indole-3-acetate, TRC

Product Code. 30381203
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30381203 10 g 10g
30377520 25 g 25g
30373790 5 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30381203 Supplier Toronto Research Chemicals Supplier No. TRCE91998010G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 778-82-5
Molecular Formula C12 H13 N O2
Molecular Weight (g/mol) 203.24
Synonym (1H-Indol-3-yl)acetic Acid Ethyl Ester, 3-[(Ethoxycarbonyl)methyl]indole, Ethyl (1H-Indol-3-yl)acetate, Ethyl 1H-Indole-3-acetate, Ethyl 2-(1H-Indol-3-yl)acetate, Ethyl 2-(Indol-3-yl)acetate, Ethyl Indol-3-ylacetate, Ethyl β-Indolylacetate, IAA Ethyl Ester, NSC 38002
IUPAC Name ethyl 2-(1H-indol-3-yl)acetate
SMILES CCOC(=O)Cc1c[nH]c2ccccc12

Specifications

Quantity 10 g
InChI Formula InChI=1S/C12H13NO2/c1-2-15-12(14)7-9-8-13-11-6-4-3-5-10(9)11/h3-6,8,13H,2,7H2,1H3
Formula Weight 203.0946
Chemical Name or Material 3-Indolyl-acetic acid-ethyl ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.