missing translation for 'onlineSavingsMsg'
Learn More

Ethyl indole-3-acetate, 98+%

Product Code. 11495543
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11495543 25 g 25g
11485543 5 g 5g
11405553 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11495543 Supplier Thermo Scientific Alfa Aesar Supplier No. A13474.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 778-82-5 | C12H13NO2 | 203.241 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 778-82-5
Molecular Formula C12H13NO2
Molecular Weight (g/mol) 203.241
MDL Number MFCD00005635
InChI Key HUDBDWIQSIGUDI-UHFFFAOYSA-N
Synonym ethyl 3-indoleacetate, ethyl 2-1h-indol-3-yl acetate, indole-3-acetic acid ethyl ester, ethyl indole-3-acetate, ethyl indol-3-ylacetate, 1h-indole-3-acetic acid, ethyl ester, ethyl 1h-indol-3-ylacetate, indole-3-acetic acid, ethyl ester, ethyl beta-indolylacetate, ethyl3-indoleacetate
PubChem CID 13067
IUPAC Name ethyl 2-(1H-indol-3-yl)acetate
SMILES CCOC(=O)CC1=CNC2=CC=CC=C21

Specifications

Melting Point 41°C to 44°C
Boiling Point 164°C to 166°C (1 mmHg)
Flash Point >110°C (230°F)
Quantity 25 g
Beilstein 171682
Formula Weight 203.24
Percent Purity ≥98%
Chemical Name or Material Ethyl indole-3-acetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.