missing translation for 'onlineSavingsMsg'
Learn More

Ethyl imidazole-4-carboxylate, 98%

Product Code. 15441139
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15441139 5 g 5g
15451139 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15441139 Supplier Thermo Scientific Alfa Aesar Supplier No. H64095.06
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 23785-21-9 | C6H8N2O2 | 140.14 g/mol

A ring-substituted-1H-imidazole-4-carboxylic acid derivative, that is shown to act as a new class of anti-tuberculosis agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A ring-substituted-1H-imidazole-4-carboxylic acid derivative, that is shown to act as a new class of anti-tuberculosis agents.

Solubility
Soluble in dimethyl sulfoxide and methanol. Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23785-21-9
Molecular Formula C6H8N2O2
Molecular Weight (g/mol) 140.14
MDL Number MFCD02646639,MFCD07363812
InChI Key KLWYPRNPRNPORS-UHFFFAOYSA-N
Synonym ethyl 1h-imidazole-4-carboxylate, ethyl imidazole-4-carboxylate, ethyl 4-imidazolecarboxylate, ethyl 4 5-imidazolecarboxylate, 4-carbethoxyimidazole, 1h-imidazole-4-carboxylic acid ethyl ester, nic7lr0zon, unii-nic7lr0zon, imidazole-4-carboxylic acid ethyl ester, 1h-imidazole-4-carboxylic acid, ethyl ester
PubChem CID 99170
IUPAC Name ethyl 1H-imidazole-5-carboxylate
SMILES CCOC(=O)C1=CN=CN1

Specifications

Melting Point 156°C to 160°C
Quantity 5 g
Beilstein 606937
Solubility Information Soluble in dimethyl sulfoxide and methanol. Slightly soluble in water.
Formula Weight 140.14
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Ethyl imidazole-4-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.