missing translation for 'onlineSavingsMsg'
Learn More

Ethyl imidazole-2-carboxylate, 97%

Product Code. 11398696
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11398696 1 g 1g
11308706 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11398696 Supplier Thermo Scientific Alfa Aesar Supplier No. L19405.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 33543-78-1 | C6H8N2O2 | 140.142 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33543-78-1
Molecular Formula C6H8N2O2
Molecular Weight (g/mol) 140.142
MDL Number MFCD03426031
InChI Key UHYNYIGCGVDBTC-UHFFFAOYSA-N
Synonym ethyl imidazole-2-carboxylate, 1h-imidazole-2-carboxylic acid, ethyl ester, ethyl1h-imidazole-2-carboxylate, 1h-imidazole-2-carboxylic acid ethyl ester, ethyl 2-imidazolecarboxylate, imidazole-2-carboxylic acid ethyl ester, 2-ethoxycarbonyl-1h-imidazole, ethylimidazole-2-carboxylate, ethoxyformyl imidazole, zlchem 847
PubChem CID 549404
IUPAC Name ethyl 1H-imidazole-2-carboxylate
SMILES CCOC(=O)C1=NC=CN1

Specifications

Melting Point 176°C to 178°C
Boiling Point 48°C to 51°C (0.3 mmHg)
Quantity 1 g
Beilstein 115685
Formula Weight 140.14
Percent Purity 97%
Chemical Name or Material Ethyl imidazole-2-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.