missing translation for 'onlineSavingsMsg'
Learn More

Ethyl hydrogen succinate, 95%

Product Code. 15491268
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15491268 25 g 25g
15471268 1 g 1g
15481268 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15491268 Supplier Thermo Scientific Alfa Aesar Supplier No. H55116.14
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 1070-34-4 | C6H10O4 | 146.142 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1070-34-4
Molecular Formula C6H10O4
Molecular Weight (g/mol) 146.142
MDL Number MFCD00053346
InChI Key LOLKAJARZKDJTD-UHFFFAOYSA-N
Synonym monoethyl succinate, ethyl hydrogen succinate, mono-ethyl succinate, butanedioic acid, monoethyl ester, butanedioic acid, 1-ethyl ester, succinic acid monoethyl ester, monoethyl butanedioate, bernsteinsaureathylester, succinic acid monoethyl, succinic acid ethyl ester
PubChem CID 70610
IUPAC Name 4-ethoxy-4-oxobutanoic acid
SMILES CCOC(=O)CCC(=O)O

Specifications

Density 1.141
Boiling Point 248°C
Flash Point 113°C (235°F)
Linear Formula HOOCCH2CH2CO2CH2CH3
Refractive Index 1.432
Quantity 25 g
Formula Weight 146.14
Percent Purity 95%
Chemical Name or Material Ethyl hydrogen succinate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.