Få mere at vide
Ethyl hydrogen pimelate, 98%
Beskrivelse
Ethyl hydrogen pimelate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ethyl hydrogen pimelate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
Solubility
Not miscible or difficult to mix with water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Tekniske data
Tekniske data
| Melting Point | 10°C |
| Density | 1.06 |
| Boiling Point | 135°C to 138°C (1 mmHg) |
| Flash Point | >110°C (230°F) |
| Linear Formula | CH3CH2O2C(CH2)5CO2H |
| Refractive Index | 1.442 |
| Quantity | 25 g |
| Beilstein | 1774583 |
| Solubility Information | Not miscible or difficult to mix with water. |
| Formula Weight | 188.22 |
| Vis mere |
RUO – Research Use Only
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.