missing translation for 'onlineSavingsMsg'
Learn More

Ethyl hydrogen pimelate, 98%

Product Code. 11366756
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11366756 5 g 5g
11376756 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11366756 Supplier Thermo Scientific Alfa Aesar Supplier No. L04025.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 33018-91-6 | C9H16O4 | 188.223 g/mol

Ethyl hydrogen pimelate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl hydrogen pimelate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Not miscible or difficult to mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33018-91-6
Molecular Formula C9H16O4
Molecular Weight (g/mol) 188.223
MDL Number MFCD00801202
InChI Key NQYXFXWKKYGBNL-UHFFFAOYSA-N
Synonym ethyl hydrogen pimelate, pimelic acid monoethyl ester, heptanedioic acid monoethyl ester, ethyl hydrogen heptane-1,7-dioate, ethyl hydroden pimelate, heptandioic acid monoethyl ester, heptanedioic acid 1-ethyl ester, heptanedioic acid, 1-ethyl ester
PubChem CID 118383
IUPAC Name 7-ethoxy-7-oxoheptanoic acid
SMILES CCOC(=O)CCCCCC(=O)O

Specifications

Melting Point 10°C
Density 1.06
Boiling Point 135°C to 138°C (1 mmHg)
Flash Point >110°C (230°F)
Linear Formula CH3CH2O2C(CH2)5CO2H
Refractive Index 1.442
Quantity 5 g
Beilstein 1774583
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 188.22
Percent Purity 98%
Chemical Name or Material Ethyl hydrogen pimelate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.