missing translation for 'onlineSavingsMsg'
Learn More

Ethyl hydrogen malonate, 90+%

Product Code. p-7059486
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11409910 5 g 5g
11419910 25 g 25g
11429910 100 g 100g
11439910 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11409910 Supplier Thermo Scientific Alfa Aesar Supplier No. 044692.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1071-46-1 | C5H8O4 | 132.115 g/mol

Ethyl hydrogen malonate is used as a reactant for the preparation of tetramic acids through Dieckmann ring closure and organocatalytic decarboxylative Doebner-Knoevenagel reactions. It is involved in the acylation reactions and Knoevenagel condensation with aldehydes. It is also used in the preparation of gamma-lactones from olefins by intermolecular carbolactonization in presence of Mn(III) acetate as a catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1071-46-1
Molecular Formula C5H8O4
Molecular Weight (g/mol) 132.115
MDL Number MFCD00020490
InChI Key HGINADPHJQTSKN-UHFFFAOYSA-N
Synonym ethyl hydrogen malonate, monoethyl malonate, monoethyl malonic acid, mono-ethyl malonate, monoethyl hydrogen malonate, malonic acid monoethyl ester, ethoxycarbonyl acetic acid, 2-ethoxycarbonyl acetic acid, 3-ethoxy-3-oxo-propanoic acid, ethylhydrogenmalonate
PubChem CID 70615
ChEBI CHEBI:86907
IUPAC Name 3-ethoxy-3-oxopropanoic acid
SMILES CCOC(=O)CC(=O)O

Specifications

Color Yellow
Density 1.119 g/mL
Boiling Point 106°C (3 mmHg)
Flash Point >110°C (230°F)
Linear Formula HO2CCH2CO2CH2CH3
Refractive Index 1.4295
Quantity 5 g
Beilstein 1758845
Solubility Information Miscible with water,chloroform and other solvents.
Formula Weight 132.12
Percent Purity ≥90%
Physical Form Liquid
Chemical Name or Material Ethyl hydrogen malonate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.