missing translation for 'onlineSavingsMsg'
Learn More

Ethyl hippurate, 96%

Product Code. p-7027761
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11456486

Brand: Thermo Scientific Alfa Aesar B24765.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1499-53-2 | C11H13NO3 | 207.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1499-53-2
Molecular Formula C11H13NO3
Molecular Weight (g/mol) 207.23
MDL Number MFCD00026890
InChI Key PTXRQIPIELXJFH-UHFFFAOYSA-N
Synonym ethyl hippurate, ethyl n-benzoylglycinate, n-benzoylglycine ethyl ester, ethyl 2-phenylformamido acetate, benzoic amide, n-ethoxycarbonyl methyl, ethylhippurate, ethyl benzoylaminoacetate, acmc-1buir, hippuric acid ethyl ester, maybridge1_006087
PubChem CID 226558
IUPAC Name ethyl 2-benzamidoacetate
SMILES CCOC(=O)CNC(=O)C1=CC=CC=C1

Specifications

Melting Point 58°C to 60°C
Quantity 5 g
Formula Weight 207.23
Percent Purity 98%
Chemical Name or Material Ethyl hippurate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.