missing translation for 'onlineSavingsMsg'
Learn More

Ethyl hexanoate, 99%

Product Code. p-7021586
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
This item is not returnable. View return policy

Product Code. 11431748

Brand: Thermo Scientific Alfa Aesar A13985.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 123-66-0 | C8H16O2 | 144.214 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-66-0
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.214
MDL Number MFCD00009511
InChI Key SHZIWNPUGXLXDT-UHFFFAOYSA-N
Synonym ethyl caproate, ethyl hexoate, hexanoic acid, ethyl ester, hexanoic acid ethyl ester, capronic ether absolute, caproic acid ethyl ester, ethyl n-hexanoate, ethyl butyl acetate, caproate, acetic acid, butyl-, ethyl ester
PubChem CID 31265
ChEBI CHEBI:86055
IUPAC Name ethyl hexanoate
SMILES CCCCCC(=O)OCC

Specifications

Melting Point -67°C
Density 0.873
Boiling Point 166°C
Flash Point 49°C (121°F)
Refractive Index 1.4075
Quantity 100 g
UN Number UN3272
Merck Index 14,3777
Formula Weight 144.21
Percent Purity 99%
Chemical Name or Material Ethyl hexanoate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.